Learn More
Lenz Laborglasinstrumente™ Erlenmeyer Flasks With Conical Ground Joint
Description
- Acc. to DIN EN ISO 4797
- With scale (approx. volume/mL)
- With inscription label
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
| Capacity (Metric) | 300 mL |
| Material | Borosilicate Glass, Duran Glass |
| Includes | With scale (approx. volume/mL), with inscription label |
| Standard Taper Joint | NS29/32 |
| Diameter (Metric) Outer | 87 mm |