Learn More
DWK Life Sciences DURAN™ Culture Tube, with straight rim, for Kapsenberg caps
Product Code. 15222079 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric):
16 mm
18 mm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
15222079 18 mm Pack of 100
11786638 16 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15222079 Supplier DWK Life Sciences Supplier No. 261322305
  • ONLINE EXCLUSIVE PRICE
    147.00 EUR
    131.46 EUR / Pack of 100
    Save 15.54 EUR
    10% Off
View all promotions
Estimated Shipment: 10-11-2026
Culture tubes are used for growing cultures from liquid media.
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today's research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used. The original DURAN™ culture tubes are distinguished by their uniform wall thickness distribution, minimal thermal expansion and the resulting high resistance to temperature changes. They are therefore particularly suitable for work at very high temperatures and rapid temperature changes.
  • Culture tubes with straight edges can be closed with metal caps or cotton plugs to allow air exchange
  • The straight rim permits the use of Kapsenberg and metal caps
Specifications
Description 1.0 to 1.2 mm thickness
Capacity (Metric) 30 mL
Material Borosilicate Glass (3.3)
For Use With Growth of microorganisms; For microbiological work
Quantity 100/Pk.
Autoclavable Autoclavable
Diameter (Metric) 18 mm