Learn More
DWK Life Sciences DURAN™ Blank for Evaporating Flask, Pear-shape
Product Code. 15201639 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15201639 1000 mL Pack of 10
10243401 2000 mL Pack of 10
15281629 250 mL Pack of 10
15291629 500 mL Pack of 10
15271629 100 mL Pack of 10
11649848 3000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15201639 Supplier DWK Life Sciences Supplier No. 210285403
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
For further processing as ground piston.
DURAN™ evaporating flasks have a uniform wall thickness and good rotational symmetry. For later use in rotary evaporators for the evaporation of solvents, a standard ground joint sleeve is formed or prepared.
  • Pear-shaped
  • Evaporating flasks have a uniform wall thickness and good rotational symmetry
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Spécification
Description Autoclavable
Material Borosilicate Glass (3.3)
For Use With (Application) For further processing as ground piston; Rotary evaporator; After further processing it can be used for evaporation of solvents
Quantity 10/Pk.
Volume (Metric) 1000 mL
Diameter (Metric) 126 mm
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.