Learn More
Lenz Laborglasinstrumente™ Duran™ Allihn Condensers with Cone
Product Code. 10306094
Change view
Click to view available options
Includes:
100 - 250 mL Extractor
70mL Extractor
DURAN™ Olive Tubing
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
10306094 70mL Extractor Each
10441064 100 - 250 mL Extractor Each
10090201 DURAN™ Olive Tubing Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10306094 Supplier Lenz Laborglasinstrumente™ Supplier No. 05226014
  • ONLINE EXCLUSIVE PRICE
    125.00 EUR
    91.51 EUR / Pack of 1
    Save 33.49 EUR
    26% Off
Estimated Shipment: 28-01-2027
Specifications
For Use With (Application) For Condensing Vapors
Includes 70mL Extractor
Quantity 1 Pack
Standard Taper Joint No NS 29/32
Material Duran Glass
Type Allihn Condenser