Learn More
Lenz Laborglasinstrumente™ Condensers Acc. to Allihn
Product Code. 10587311
Change view
Click to view available options
Standard Taper Joint No:
14/23
19/26
29/32
Length (Metric) Jacket:
160 mm
250 mm
400 mm
500 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Jacket unitSize
10587311 14/23 250 mm Each
10743605 29/32 160 mm Each
10419942 19/26 250 mm Each
10324902 14/23 160 mm Each
10151502 29/32 250 mm Each
10755013 29/32 400 mm Each
11856074 29/32 500 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10587311 Supplier Lenz Laborglasinstrumente™ Supplier No. 05223301
  • ONLINE EXCLUSIVE PRICE
    101.00 EUR
    80.52 EUR / Pack of 1
    Save 20.48 EUR
    20% Off
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
Facilitates the condensing of vapors in the laboratory. Lenz Laborglasinstrumente™ Condensers Acc. to Allihn are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN 12581
  • With NS socket and cone
  • Corrosion-free and resistant to thermal shock
  • Available with either glass olive or GL plastic olives/threads
Specifications
For Use With (Application) For Condensing Vapors
Length (Metric) Jacket 250 mm
Quantity 1 Pack
Standard Taper Joint No 14/23
Material Duran Glass
Type Allihn Condenser