Learn More
Lenz Laborglasinstrumente™ Two-Neck Adapters
Description
- Acc. to DIN 12594
- Two NS sockets and one NS cone
- High taper accuracy of the cone
- Minimized need for grease
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Material | Duran |
| Type | 2-neck Adapter |
| Features | With two parallel necks, with two NS sockets and one NS cone |