Learn More
R&D Systems™ DuoSet ELISA Ancillary Reagent Kit 2
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit (5 x 384-well Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |