Learn More
PureAire Monitoring Systems, Inc. Dual O2/CO2 (0-25% O2, 0-50,000ppm CO2) with 10+ Year Sensors
Product Code. 16228831
Change view
Click to view available options
Certifications/Compliance:
CE
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Certifications/Compliance Unit Size
16228831 CE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16228831 Supplier PureAire Monitoring Systems, Inc. Supplier No. 99144
  • ONLINE EXCLUSIVE PRICE
    3600.00 EUR
    2602.65 EUR / Pack of 1
    Save 997.35 EUR
    27% Off
Estimated Shipment: 23-12-2026
Perfect for your facility if you use nitrogen, helium, and/or argon gases. The monitor is also suited for use in confined spaces such as cryogenic facilities. Sensors lasting 10 years and more.
PureAire Monitoring System’s Dual O2/CO2 Monitor provides continuous monitoring of oxygen and carbon dioxide. This monitor is ideal for use in spaces where low oxygen concentrations could pose a health hazard, from nitrogen gas storage sites to CO2 beverage gas distribution areas and more.
  • No maintenance zirconium O2 cell and long life CO2 sensor
  • No calibration required for oxygen Self-calibrating CO2 sensor
  • Local backlit concentration display
  • Built-in audible horn
  • Built-in visual LED alarm indicators
  • Alarm level 1 and 2 for O2 and CO2
  • No drift due to environmental or temperature changes
  • Plug-in the wall power supply included- 24V, 220V
What is a Dual O2/CO2 Monitor?
  • A Dual O2/CO2 Monitor is a stand-alone system ideal for the continuous monitoring of nitrogen or cryogenics near freezers, confined spaces, and other locations where low oxygen levels may pose a hazard to personnel.
Why choose PureAire?
  • As an industry leader in oxygen monitoring and gas detection, PureAire is dedicated to the safety of our clients, and the customers they serve, by manufacturing monitors that are long-lasting, innovative, and reliable.
What gases can a Dual O2/CO2 Monitor be used for?
  • PureAire’s oxygen monitors can be used for CO2, nitrogen, argon, and helium gases.
Installation is easy. Plug-in ready.
  • The monitor will arrive ready to work right out of the box. Simply secure the unit on the wall 1-2 meters from the gas source, plug it in, and power it on. The monitor will begin sampling oxygen levels after a 4 minute startup period. Levels are displayed clearly on the screen for easy access and visibility.  
Certifications
CE Marked
Warranty
2 years
Specifications
Certifications/Compliance CE
Depth (English) 3 in.
Depth (Metric) 76 mm
Dimensions (L x W x H) 3 x 6.5 x 3.15 in.
Height (English) 3.15 in.
Height (Metric) 80 mm
Humidity Range 0-95% RH
Temperature Range (English) -40°F to +134°F
Temperature Range (Metric) -40 to +55°C
Voltage 24V
Warranty 2 Year after ETD
Weight (English) 1.1 lb.
Weight (Metric) 0.5 kg
Width (English) 6.5 in.
Width (Metric) 165 mm