Learn More
Fisherbrand™ Dual LCD Display Thermo-Hygrometer
Product Code. 11945148
Change view
Click to view available options
Temperature Range (Metric):
-10°C to +60°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
11945148 -10°C to +60°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11945148 Supplier Fisherbrand™ Supplier No. 30.5002
  • ONLINE EXCLUSIVE PRICE
    24.12 EUR
    17.71 EUR / Pack of 1
    Save 6.41 EUR
    26% Off
Estimated Shipment: 07-12-2026
Specifications
Type Thermohygrometer
Humidity Range 10 to 99%
Resolution 0.01
Temperature Range (Metric) -10°C to +60°C
Display Type Dual LCD
Length (Metric) 110 mm
Height (Metric) 110 mm
Voltage 1.5 V
Includes 2-AAA Batteries