Learn More
KGW Isotherm™ DSS Stainless Steel Dewar Flasks
Product Code. 10722023
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
3 L
Height (Metric):
178 mm
190 mm
225 mm
305 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) unitSize
10722023 1 L 225 mm Each
10065171 1 L 178 mm Each
10594502 2 L 305 mm Each
10228511 3 L 190 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10722023 Supplier KGW Isotherm™ Supplier No. 2101
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Lid is sold separately; see KGW Isotherm Lids (10166941).
Specifications
For Use With (Application) Used to Store and Transport liquid nitrogen ( LN2 ), Dry Ice ( CO2 ) and Other Coolants
Height (Metric) 225 mm
Capacity (Metric) 1 L
Diameter (Metric) Inner 85 mm
Material DSS Stainless Steel