Learn More
Heinz Herenz Dry Swabs
Product Code. 15283728
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 150
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
15283728 Sterile Pack of 150
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15283728 Supplier Heinz Herenz Supplier No. 1020005
  • ONLINE EXCLUSIVE PRICE
    83.59 EUR
    50.51 EUR / Pack of 150
    Save 33.08 EUR
    39% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Dry swabs with medium/ transport swabs. Sterile with Amies medium. Plastic applicator with viscose/rayon head, sterilized by radiation.
We offer a wide range of sterile and nonsterile swabs:
  • Wood, PS, PP, aluminium or paper applicator
  • Each in various lengths available
  • With cotton, viscose, polyester or foam head
  • Diameter of the heads: small, medium or large
  • This guarantees that the right instrument is available for every special application.
  • For example, the cotton-tipped swabs are recommended for use by ENT specialists.
  • Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
  • Only suitable for single use
  • Use by medical personnel only
Specifications
Product Type Applicator Swab and Tube, Swab
Format Swab
Media Type Liquid Amies, Amies Medium
Sterility Sterile
Color White
Quantity 150Pack
Applicator Material Polystyrene
Tip Type Viscose
Material Plastic
Tip Material Polystyrene