Learn More
Welch Ilmvac™ Diaphragm Vacuum Pump Service and Maintenance Kit
Product Code. 10245242 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With:
MP/MPC 101 Z, 201 E
MP/MPC 201 T, MP 101V
MP/MPC 301 Z, 601 E, 301 Z
MP/MPC 601 T, 901 Z, 1201 E, MP 301V
MP/MPR 060 E, 030 Z, Biovac 060, MP 101V
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
10245242 MP/MPC 301 Z, 601 E, 301 Z Each
13191984 MP/MPC 101 Z, 201 E Each
10402742 MP/MPC 201 T, MP 101V Each
10090972 MP/MPC 601 T, 901 Z, 1201 E, MP 301V Each
10328687 MP/MPR 060 E, 030 Z, Biovac 060, MP 101V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10245242 Supplier Welch Ilmvac™ Supplier No. 402041
  • ONLINE EXCLUSIVE PRICE
    262.00 EUR
    207.30 EUR / Pack of 1
    Save 54.70 EUR
    20% Off
Estimated Shipment: 22-12-2026
Service and Maintenance Kits comprised of replacement seals and wearable parts for various models of diaphragm pumps.
  • Available for a range of diaphragm vacuum pump models
  • Provided individually
TRUSTED_SUSTAINABILITY
Specifications
For Use With MP/MPC 301 Z, 601 E, 301 Z
Includes Diaphragm, Valves, and O-rings
Quantity 1 Ea.
Type Service Kit