Learn More
Kimble Chase™ Drummond Microcaps Borosilicate Glass Capillary Tubes
Description
Kimble Chase Drummond Microcaps Borosilicate Glass Capillary Tubes, Capacity: 0.5 uL, Length: 32 mm, Quantity: 100/Pk.
Specifications
Specifications
| Capacity (Metric) | 50 μL |
| Material | Borosilicate Glass |
| Quantity | 100/Pk. |
| Type | Capillary Tube |
| Length (Metric) | 100 mm |