Learn More
Kimble Chase™ Drummond Microcaps Borosilicate Glass Capillary Tubes
Product Code. 13035813
Change view
Click to view available options
Capacity (Metric):
0.5 μL
1 μL
25 μL
50 μL
Length (Metric):
32 mm
65 mm
100 mm
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Unit Size
13035813 25 μL 65 mm Pack of 100
13065803 0.5 μL 32 mm Pack of 100
12389389 1 μL - Pack of 100
13045813 50 μL 100 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 13035813 Supplier Kimble Chase™ Supplier No. 7645000025
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
 Restricted Item
Estimated Shipment: 10-12-2026
Kimble Chase Drummond Microcaps Borosilicate Glass Capillary Tubes, Capacity: 0.5 uL, Length: 32 mm, Quantity: 100/Pk.
Specifications
Capacity (Metric) 25 μL
Material Borosilicate Glass
Quantity 100/Pk.
Type Capillary Tube
Length (Metric) 65 mm