Learn More
Drum Key, Supelco™
Product Code. 15855180
Change view
Click to view available options
Quantity:
1 U
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15855180 1 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15855180 Supplier Supelco Supplier No. 1088030001
 Restricted Item
Estimated Shipment: 09-11-2026
For use with containers with 2 in. and 3/4 in. screw caps
for opening and closing containers with 2\ and 3/4\ screw caps
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 U