Learn More
MP Biomedicals Drug of Abuse Rapid Test - Cup 13 parameters (AMP1000/BAR300/BUP10/BZD300/COC300/EDDP100/K2-SPICE50/M-AMP1000/MDMA500/OPI300/TCA1000/THC50/CREA)
Shop All MP Biomedicals ProductsDescription
The Drug of Abuse Rapid Test - Cup is based on the principle of specific immunochemical reaction between antibodies and antigen to analyze particular compounds in human urine specimen. The assay relies on the competition for the binding antibody. When drug is present in the urine specimen, it competes with drug conjugate for the limited amount of antibody-dye conjugate. When the amount of drug is equal or more than the cut-off, it will prevent the binding of drug conjugate to the antibody. Therefore, a positive urine specimen will not show a colored band on the test line zone, indicating a positive result, while the presence of a colored band indicates a negative result. A control line is present in the test window to work as procedural control. This colored band should always appear on the control line zone if the test device is stored in good condition and the test is performed appropriately.
- CE-marked
- Drug detection in human urine sample
- Tailor-made tests with customized combinations
- More than 30 parameters: amphetamine, benzodiazepine, barbiturate, buprenorphine, cocaine, EDDP, ketamine, methamphetamine, MDMA, methadone, opiates, oxycodone, pencyclidine, THC, propoxyphene, tramadol, tricyclic antidepressant, methylphenidate, fentanyl, synthetic cannabis, clonazepam, cotinine, alcohol Adulteration parameters: oxidants, bleach, pyridinium chlorochromate, nitrite, creatinine, pH and specific gravity
- For this reference, the parameters and cut-off values are: Amphetamine 1000 ng/mL, Barbiturate 300 ng/mL, Buprenorphine 10 ng/mL, Benzodiazepine 300 ng/mL, Cocaine 300 ng/mL, EDDP 100 ng/mL, Synthetic Cannabis 50 ng/mL, Methamphetamine 1000 ng/mL, MDMA 500 ng/mL, Opiate 300 ng/mL, Tricyclic Antidepressant 1000 ng/mL, THC 50 ng/mL, and Creatinine.
Spécification
Spécification
| CE Marker | Self-declared |
| Clia Complexity | Not Applicable |
| Detectable Analytes | Amphetamine (AMP), Barbiturates (BAR), Buprenorphine (BUP), Benzodiazepines (BZO), Cocaine (COC), Methadone Metabolite (EDDP), Synthetic Marijuana (K2), Methamphetamine (mAMP), Methylenedioxymethamphetamine (MDMA), Opiates (OPI), Tricyclic Antidepressants (TCA), Cannabinoids (THC), Creatinine |
| Form | Ready-to-Use (RTU) |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.