Learn More
Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped, with PTFE plug
Product Code. 11886184
Change view
Click to view available options
Capacity (Metric):
100 mL
1000 mL
2000 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11886184 100 mL Each
11826214 1000 mL Each
11896144 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11886184 Supplier Lenz Laborglasinstrumente™ Supplier No. 04473702
  • ONLINE EXCLUSIVE PRICE
    94.00 EUR
    72.68 EUR / Pack of 1
    Save 21.32 EUR
    22% Off
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Made of borosilicate glass. Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped, with PTFE plug are used for transferring fluids. The funnel is fitted with a stopcock which allows the flow to be controlled.
  • Conical shape with NS sleeve and NS core
  • With PTFE plug
  • With threaded valve protection
  • Borosilicate glass
Specifica
Capacity (Metric) 100 mL
Material Borosilicate Glass
Pore Size 2.5 mm bore
Stopper Material PTFE
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.