Learn More
Drometrizole Trisiloxane, TRC
Product Code. 30134096
Change view
Click to view available options
Quantity:
100 mg
200 mg
1 g
Unit Size:
100mg
1g
200mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134096 1 g 1g
30006964 100 mg 100mg
30005824 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134096 Supplier Toronto Research Chemicals Supplier No. TRCD6794101G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 155633-54-8
Molecular Formula C24 H39 N3 O3 Si3
Molecular Weight (g/mol) 501.84
Synonym Phenol, 2-(2H-benzotriazol-2-yl)-4-methyl-6-[2-methyl-3-[1,3,3,3-tetramethyl-1-[(trimethylsilyl)oxy]disiloxanyl]propyl]- (9CI), 2-(2H-Benzotriazol-2-yl)-4-methyl-6-[2-methyl-3-[1,3,3,3-tetramethyl-1-[(trimethylsilyl)oxy]-1-disiloxanyl]propyl]phenol, 2-(2H-Benzotriazol-2-yl)-4-methyl-6-[2-methyl-3-[1,3,3,3-tetramethyl-1-[(trimethylsilyl)oxy]disiloxanyl]propyl]phenol, Drometrizole trisiloxane, Mexoryl XL, OR 10154, Silatrizole
IUPAC Name 2-(benzotriazol-2-yl)-4-methyl-6-[2-methyl-3-[methyl-bis(trimethylsilyloxy)silyl]propyl]phenol
SMILES CC(Cc1cc(C)cc(c1O)n2nc3ccccc3n2)C[Si](C)(O[Si](C)(C)C)O[Si](C)(C)C
Spezifikation
Quantity 1 g
InChI Formula InChI=1S/C24H39N3O3Si3/c1-18-14-20(15-19(2)17-33(9,29-31(3,4)5)30-32(6,7)8)24(28)23(16-18)27-25-21-12-10-11-13-22(21)26-27/h10-14,16,19,28H,15,17H2,1-9H3
Formula Weight 501.2299
Percent Purity >95%
Chemical Name or Material Drometrizole Trisiloxane 2-(2H-Benzotriazol-2-yl)-6-{3-[bis(trimethylsiloxy)methylsilyl]isobutyl}-p-cresol
Grade HPLC