Learn More
Buerkle™ Drill Bit for Buerkle™ Mole
Product Code. 10391224 Shop All Bürkle Products
Change view
Click to view available options
For Use With (Application):
For Clay and Wet Soils
For Coarse Sand
For Collecting and Removing Stones
For Combination Soils
For Gravel
For Riverbank Soil Horizons
For Sand
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
10391224 For Clay and Wet Soils Each
10095713 For Coarse Sand Each
10107503 For Collecting and Removing Stones Each
10571925 For Combination Soils Each
10637674 For Gravel Each
10035673 For Sand Each
10443015 For Riverbank Soil Horizons Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10391224 Supplier Buerkle™ Supplier No. 53501015
Control Your Critical Environments
Use promo code 28022
View all promotions
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Buerkle™ Drill Bit for Buerkle™ Mole are specially designed for very specific soil properties.
  • Clay and damp soil
    • With narrow side panels for minimum soil friction/resistance. As wet soils are usually very cohesive, the soil still adheres to the bit very well.
  • Sand
    • With broad side panels and therefore a large retention capacity for the sand.
  • Coarse sand
    • With extra broad side panels enclosing even more of the bit so that even coarse sandy soils and very dry soils are retained.
  • Combination
    • Retains both strongly cohesive and less cohesive soils.
  • Riverside
    • Very suitable for drilling in hard, encrusted soils and soils mixed with fine gravel. The drill cutters project from the drill itself, so that friction is greatly reduced.
  • Gravel drill
    • With slightly rounded, oblique scoops which pull themselves downward in the gravel. Any stones present are clamped between the drill sides. Finer soil material (coarse sand with fine gravel) is retained in the drill bit.
  • Stone catcher
    • The stone catcher is used to collect and remove stones that prevent proper sampling with one of the other drill bits.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Drill Bit
Description Equipment for mole drill sampler
Includes Narrow Side Panels
For Use With (Application) For Clay and Wet Soils
For Use With (Equipment) Mole Drill Sampler