Learn More
Lenz Laborglasintrumente™ Drechsel Gas Washing Bottles
Product Code. 10450323
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Porosity:
1
2
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Porosity Unit Size
10450323 500 mL 2 Each
11536924 500 mL 1 Each
10622092 1000 mL 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10450323 Supplier Lenz Laborglasintrumente™ Supplier No. 05510258
  • ONLINE EXCLUSIVE PRICE
    112.00 EUR
    87.62 EUR / Pack of 1
    Save 24.38 EUR
    21% Off
Estimated Shipment: 30-12-2026
Gas washing bottles acc. to Drechsel with filter disc
  • Acc. to DIN 12596
  • Complete with head (with filter disc)
  • Ground joint connection: NS 29/32
  • Material: DURAN™ tubing (head), borosilicate glass 3.3 (bottle)
Specifications
Capacity (Metric) 500 mL
Type Olive Connections
Includes DURAN™ tubing head, borosilicate glass 3.3 bottle
Porosity 2