Learn More
Hirschmann™ Automatic Burettes, Acc. to Dr. Schilling
Product Code. 10360001
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
15 mL
25 mL
50 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10360001 50 mL Each
10195920 10 mL Each
10569222 15 mL Each
10441331 25 mL Each
10350411 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10360001 Supplier Hirschmann™ Supplier No. 3600175
  • ONLINE EXCLUSIVE PRICE
    117.00 EUR
    103.00 EUR / Pack of 1
    Save 14.00 EUR
    11% Off
Estimated Shipment: 19-10-2026
Excellent for automatically dispensing measured amounts of chemical solutions in the laboratory. Hirschmann Instruments™ Automatic Burettes, Acc. to Dr. Schilling are available in a range of capacities to fit your needs.
  • With Schellbach stripe
  • Blue graduation
  • Fast flow burette
Specifications
Capacity (Metric) 50 mL
Material Glass
Graduations 0.1 mL
Includes LDPE Bottle
Scale Blue
Tolerance ±0.1 mL
Type Schilling Buret
Certifications/Compliance DKD Certificate