Learn More
Dräger™ Sulfur Dioxide Replacement Sensor
Product Code. 15179992
Change view
Click to view available options
Measurement Range:
0 to 50
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
15179992 0 to 50 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15179992 Supplier Dräger™ Supplier No. 6809160
  • ONLINE EXCLUSIVE PRICE
    1304.63 EUR
    1083.72 EUR / Pack of 1
    Save 220.91 EUR
    16% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Sensor for use with X-am 7000 Multi-Gas Monitors
  • Detection Range: 0-50
  • Detects: SO2
Warranty
1 Year
Specifications
Components Measured Sulfur Dioxide (XS)
Humidity Range 10 to 90%
Measurement Range 0 to 50
Pressure Range 700 to 1300mbar
Resolution 0.1
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40° to +50°C
Warranty 1 Year