Learn More
Dräger™ Short-Term Detector Tubes: Water Vapor
Product Code. 10296602
Change view
Click to view available options
Measurement Range:
0.05 to 1
1 to 40
20 to 1500
20 to 40mg/L
3 to 60
5 to 200
Tube Type:
Water vapor 0.1
Water vapor 0.1/a
Water vapor 1/b
Water vapor 20/a-P
Water vapor 3/a
Water vapor 5/a-p
Response Time:
1.5 min.
10 to 5 min.
2 min.
20/40 sec.
25 min.
Not Applicable
Unit Size:
Each
Pack of 10
Pack of 3
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Tube Type Response Time Unit Size
10296602 20 to 1500 Water vapor 20/a-P 10 to 5 min. Each
10318562 5 to 200 Water vapor 5/a-p 25 min. Pack of 10
15242770 3 to 60 Water vapor 3/a Not Applicable Pack of 3
10443922 0.05 to 1 Water vapor 0.1/a 1.5 min. Pack of 10
10729314 20 to 40mg/L Water vapor 1/b 20/40 sec. Pack of 10
10062541 1 to 40 Water vapor 0.1 2 min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10296602 Supplier Dräger™ Supplier No. 8103061
  • ONLINE EXCLUSIVE PRICE
    202.35 EUR
    163.48 EUR / Pack of 1
    Save 38.87 EUR
    19% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ Customer Service for more information.
Specifications
Components Measured Water Vapor
For Use With (Equipment) Aerotest System
Humidity Range Not Applicable
Measurement Range 20 to 1500
Quantity 10
Response Time 10 to 5 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Tube Type Water vapor 20/a-P