Learn More
Dräger™ Short-Term Detector Tubes - Ozone
Product Code. 10120921
Change view
Click to view available options
Components Measured:
Ozone 0.05/b
Ozone 10/a
Measurement Range:
0.05 to 0.7ppm
20 to 300ppm
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Measurement Range Unit Size
10120921 Ozone 0.05/b 0.05 to 0.7ppm Pack of 10
10355142 Ozone 10/a 20 to 300ppm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10120921 Supplier Dräger™ Supplier No. 6733181
  • ONLINE EXCLUSIVE PRICE
    258.80 EUR
    204.22 EUR / Pack of 10
    Save 54.58 EUR
    21% Off
 Restricted Item
Estimated Shipment: 14-12-2026
For making spot measurements for various gases and vapors
  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.
Specifications
Components Measured Ozone 0.05/b
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Measurement Range 0.05 to 0.7ppm
Quantity 10/Box