Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen
Product Code. 10530663
Change view
Click to view available options
Components Measured:
Hydrogen Fluoride 1.5/b (gas or vapor)
Hydrogen Peroxide 0.1/a (gas or vapor)
Measurement Range:
0.1 to 3ppm
15ppm
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Measurement Range Unit Size
10530663 Hydrogen Peroxide 0.1/a (gas or vapor) 0.1 to 3ppm Pack of 10
10325192 Hydrogen Fluoride 1.5/b (gas or vapor) 15ppm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10530663 Supplier Dräger™ Supplier No. 8101041
  • ONLINE EXCLUSIVE PRICE
    218.33 EUR
    171.27 EUR / Pack of 10
    Save 47.06 EUR
    21% Off
Estimated Shipment: 29-12-2026
For making spot measurments for various gases and vapors
  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Hydrogen Peroxide 0.1/a (gas or vapor)
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Measurement Range 0.1 to 3ppm
Quantity 10/Box