Learn More
Dräger™ Individual Haz Mat Simultaneous Test Sets
Product Code. 12702517
Change view
Click to view available options
Type:
Simultaneous Test Set I
Simultaneous Test Set II
Simultaneous Test Set III
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12702517 Simultaneous Test Set I Each
10385704 Simultaneous Test Set II Each
10345754 Simultaneous Test Set III Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
No other type of tube should be substituted with these kits.
Product Code. 12702517 Supplier Dräger™ Supplier No. 8101735
  • ONLINE EXCLUSIVE PRICE
    192.77 EUR
    157.46 EUR / Pack of 1
    Save 35.31 EUR
    18% Off
 Restricted Item
Estimated Shipment: 30-12-2026
No other type of tube should be substituted with these kits.
Requires
Require adapter and a Dräger pump (sold separately) for the semi-quantitative determination of inorganic gases and organic vapors.
For simultaneous testing of five different gases
Test Set I. Includes detector tubes for hydrogen chloride, hydrogen cyanide, carbon monoxide, ammonia, and nitrous fumes.
Test Set II. Includes detector tubes for sulfur dioxide, chlorine, hydrogen sulfide, carbon dioxide, and phosgene.
Test Set III. Includes detector tubes for alcohols, aliphatic hydrocarbons, aromatics, chlorinated hydrocarbons, and ketones.
Order Info
Packs of 10 test sets.
Specifications
Type Simultaneous Test Set I
For Use With (Application) For simultaneous testing of five different gases
Includes Tests for hydrogen chloride, hydrogen cyanide, carbon monoxide, ammonia, and nitrous fumes