Learn More
Dräger™ CMS Chips: Xylene
Product Code. 10682333
Change view
Click to view available options
Measurement Range:
10 to 300
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
10682333 10 to 300 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10682333 Supplier Dräger™ Supplier No. 6406260
  • ONLINE EXCLUSIVE PRICE
    233.24 EUR
    198.50 EUR / Pack of 1
    Save 34.74 EUR
    14% Off
Estimated Shipment: 22-12-2026
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type o-Xylene CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 10 to 300
Response Time 75 to 500 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C