Learn More
Dräger™ CMS Chips: Hydrogen Peroxide
Product Code. 12364619
Change view
Click to view available options
Measurement Range:
0.2 to 2
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
12364619 0.2 to 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12364619 Supplier Dräger™ Supplier No. 6406440
  • ONLINE EXCLUSIVE PRICE
    418.55 EUR
    358.50 EUR / Pack of 1
    Save 60.05 EUR
    14% Off
Estimated Shipment: 23-12-2026
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Hydrogen Peroxide CMS Chip
Humidity Range 1 to 20mg/L
Measurement Range 0.2 to 2
Response Time 180 to 600 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C