Learn More
Dräger™ CMS Chips: Hydrogen Cyanide
Product Code. 10719772
Change view
Click to view available options
Measurement Range:
2 to 50
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
10719772 2 to 50 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10719772 Supplier Dräger™ Supplier No. 6406100
  • ONLINE EXCLUSIVE PRICE
    288.62 EUR
    248.27 EUR / Pack of 1
    Save 40.35 EUR
    13% Off
Estimated Shipment: 12-01-2027
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Hydrogen Cyanide CMS Chip
Humidity Range 1 to 20mg/L
Measurement Range 2 to 50
Response Time 30 to 260 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 30°C