Learn More
Dräger™ 7000 Tube Opener
Product Code. 10200383
Change view
Click to view available options
For Use With (Equipment):
Detector Tube Systems
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10200383 Detector Tube Systems Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10200383 Supplier Dräger™ Supplier No. 6401200
  • ONLINE EXCLUSIVE PRICE
    101.87 EUR
    87.87 EUR / Pack of 1
    Save 14.00 EUR
    13% Off
Estimated Shipment: 11-12-2026
An accessory for detector tube systems.
  • Accessory for detector tube systems
  • For scoring and breaking tips off individual Draeger detector tubes
  • Glass tips safely stored for proper disposal
  • Aids in breaking tube ampoules
Specifications
For Use With (Equipment) Detector Tube Systems
Type 7000 Tube Opener