missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPPIV/CD26 Antibody (DPP4/910), Novus Biologicals™
Mouse Monoclonal Antibody
290.00 EUR - 509.00 EUR
Specifications
| Antigen | DPPIV/CD26 |
|---|---|
| Clone | DPP4/910 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Frozen 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18148367
|
Novus Biologicals
NBP2-44572-0.02MG |
0.02 mg |
290.00 EUR
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18129808
|
Novus Biologicals
NBP2-44572-0.1MG |
0.1 mg |
509.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence
DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Human, Rat, Porcine (Negative), Sheep (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Cell Surface), Immunofluorescence.Specifications
| DPPIV/CD26 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cellular Markers, GPCR, Immunology | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
| DPP4 | |
| IgG2b κ | |
| Protein A or G purified | |
| Recognizes a glycoprotein of 110kDa, identified as CD26. It is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. |
| DPP4/910 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Frozen 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Porcine (Negative), Sheep (Negative) | |
| P27487 | |
| 1803 | |
| Recombinant human DPP4 protein | |
| Primary | |
| Store at 4C. | |
| 110 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title