missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPPIV/CD26 Antibody (DPP4/910), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44572-0.02MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence
DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Human, Rat, Porcine (Negative), Sheep (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Cell Surface), Immunofluorescence.
Specifications
| DPPIV/CD26 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Frozen 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| P27487 | |
| DPP4 | |
| Recombinant human DPP4 protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a glycoprotein of 110kDa, identified as CD26. It is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. | |
| Store at 4C. | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence | |
| DPP4/910 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
| Mouse | |
| 110 kDa | |
| 0.02 mg | |
| Adaptive Immunity, Cellular Markers, GPCR, Immunology | |
| 1803 | |
| Human, Rat, Porcine (Negative), Sheep (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction