Learn More
DPPC, MedChemExpress
Product Code. 16480627
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480627 50 mg 50mg
16484298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16480627 Supplier Medchem Express Supplier No. HY109506
 Restricted Item
Estimated Shipment: 25-01-2027
MedChemExpress DPPC (129Y83) is a zwitterionic phosphoglyceride that can be used for the preparation of liposomal monolayers. DPPC-liposome serves effectively as a delivery vehicle for inducing immune responses against GSL antigen in mice.
DPPC (129Y83) is a zwitterionic phosphoglyceride that can be used for the preparation of liposomal monolayers. DPPC-liposome serves effectively as a delivery vehicle for inducing immune responses against GSL antigen in mice.
Specifications
Chemical Name or Material DPPC
CAS 63-89-8
Color White
Purity Grade Notes Research
Molecular Formula C40H80NO8P
Quantity 50 mg
Synonym 129Y83
Solubility Information Ethanol : 25 mg/mL (34.06 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (6.81 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CCCCCCCCCCCCCCCC(O[C@H](COC(CCCCCCCCCCCCCCC)=O)COP(OCC[N+](C)(C)C)([O-])=O)=O
Molecular Weight (g/mol) 734.04
Formula Weight 734.04
Percent Purity 98.0%
Grade Research
Physical Form Solid