Learn More
Lovibond™ DPD No.1 Tablet
Product Code. 13151681
Change view
Click to view available options
Quantity:
250/Pk.
Unit Size:
250 tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13151681 250/Pk. 250 tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13151681 Supplier Lovibond™ Supplier No. 511051BT
  • ONLINE EXCLUSIVE PRICE
    57.55 EUR
    29.04 EUR / 250 tablets
    Save 28.51 EUR
    49% Off
 Restricted Item
Estimated Shipment: 11-01-2027
A high quality comparator instrument for visually matching the intensity of sample color with Lovibond™ glass filters
Lovibond™ Comparator 2000+
A high quality comparator instrument for visually matching the intensity of sample color with Lovibond™ glass filters. It includes an integrated prism, which brings the sample and comparison fields together, allowing enhanced color discrimination and matching. The prism is ultrasonically sealed to prevent contamination of the optics. It includes the facility to compensate for water samples that are turbid or tinted.
Lovibond™ Daylight 2000 unit
A standardised bench top light source that guarantees constant and uniform lighting conditions for color matching with the Lovibond™ Comparator 2000+ instruments (comparator not included).
Lovibond™ Water Test Discs and Reagents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. Some of the more popular discs are listed here but other ranges and tests can be supplied on request (for example chlorine dioxide, copper, fluoride, hydrazine, hydrogen peroxide, iodine, manganese, molybdate, nitrite, oxygen, ozone, phosphate, QAC, silica, sulfide, zinc).
What you need
  • Comparator instrument (appropriate Nessleriser)
  • Lovibond™ disc, specific to the test
  • Reagents, specific to the test
  • Sample container of the required path length
  • Lighting unit (optional) to ensure constant viewing conditions
  • Simple and flexible system giving reliable results in both laboratory and field testing
  • Compact, easily portable and robust for field use
  • Reagents mostly available in convenient tablet form
  • Includes stable colored glass filters which can be certified under a ISO 9001 QA procedure
Specifications
Packaging 250/Pk.
Quantity 250/Pk.