Learn More
Gibco™ DPBS (10X), no calcium, no magnesium
Product Code. 11520486
Change view
Click to view available options
Quantity:
100mL
500mL
10 x 500 mL
20 x 100 mL
Unit Size:
100mL
2000mL
5000mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11520486 100mL 100mL
11530486 500mL 500mL
11550486 20 x 100 mL 2000mL
11540486 10 x 500 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11520486 Supplier Gibco™ Supplier No. 14200059
DPBS (10X), no calcium, no magnesium
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DPBS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DPBS
Concentration 10 X
For Use With (Application) Mammalian Cell Culture
pH 6 to 7
Physical Form Liquid
Quantity 100mL
Includes No Sodium Pyruvate
Osmolality 2500 to 2900 mOsm/kg