Learn More
Thermo Scientific Chemicals DuPont Molykote High-Vacuum Grease
Product Code. 15471977
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471977 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15471977 Supplier Thermo Scientific Chemicals Supplier No. 044224.KT
  • ONLINE EXCLUSIVE PRICE
    109.00 EUR
    84.65 EUR / Pack of 1
    Save 24.35 EUR
    22% Off
 Restricted Item
In Stock
Add to basket
Edge
Recommended for vacuums reaching between 1 x 10-5 and 1 x 10-6 mm HgDow Corning™ High-Vacuum Grease is used as an anti-seize grease in ground glass joints. It is used to lubricate synthetic and silicone rubber gaskets. Further, it is used to lubricate plug valves, control valves, flow meter bearings, ceramic plug cocks and fire extinguisher valves. In addition to this, it is used to lubricate water-treating equipment, laboratory equipment, lubricate O-rings on binoculars and telescopes and to prevent fogging of delicate lenses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used as an anti-seize grease in ground glass joints. Also used to lubricate synthetic and silicone rubber gaskets. Further, it is used to lubricate plug valves, control valves, flow meter bearings, ceramic plug cocks and fire extinguisher valves. In addition to this, it is used to lubricate water-treating equipment, laboratory equipment, lubricate O-rings on binoculars and telescopes and to prevent fogging of delicate lenses.
Packaging Tube
Quantity 1 Ea.
Temperature (Metric) -40°C to +204°C
Weight (English) 5.3 oz.
Weight (Metric) 150 g