Learn More
Metro™ Double-Deep qwikTRAK™ Mobile Unit Kits
Product Code. 15906991
Change view
Click to view available options
Width (English):
18 in.
21 in.
24 in.
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Heavy Polymer
Metroseal Green Epoxy
Stainless Steel
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Width (English) Shelf Finish unitSize
15906991 21 in. 21 in. Stainless Steel Each
15936991 24 in. 24 in. Stainless Steel Each
15946991 24 in. 24 in. Heavy Polymer Each
15976981 18 in. 18 in. Stainless Steel Each
15916991 24 in. 24 in. Chrome Each
15986981 18 in. 18 in. Heavy Polymer Each
15926991 24 in. 24 in. Metroseal Green Epoxy Each
15996981 21 in. 21 in. Chrome Each
15966981 18 in. 18 in. Metroseal Green Epoxy Each
15956981 18 in. 18 in. Chrome Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15906991 Supplier Metro™ Supplier No. LBTM21S
  • ONLINE EXCLUSIVE PRICE
    2015.00 EUR
    1809.39 EUR / Pack of 1
    Save 205.61 EUR
    10% Off
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
Kit provides components for shelving units that fit on tracks for added mobility
  • For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax i™ shelving systems
  • Kit includes eight 74 in. (188cm) high -UP posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Order Info
Order one kit per double-deep system. Shelves are sold separately.
Specifications
Product Type Mobile Unit Kit
Finish Stainless Steel
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ shelving system
Includes Eight 74 in. high up posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Width (English) Shelf 21 in.
Width (English) 21 in.
Width (Metric) Shelf 53 cm
Width (Metric) 53 cm
Material Stainless Steel