Learn More
DOTA, TRC
Product Code. 30012185
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012185 500 mg 500mg
30004665 250 mg 250mg
17962115 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012185 Supplier Toronto Research Chemicals Supplier No. TRCD535500500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60239-18-1
Molecular Formula C16 H28 N4 O8
Molecular Weight (g/mol) 404.42
Synonym 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, 1,4,7,10-Tetra(carboxymethyl)-1,4,7,10-tetraazacyclododecane, 1,4,7,10-Tetraazacyclodecane-1,4,7,10-tetraacetic acid, 1,4,7,10-Tetraazacyclododecane-N,N',N' ',N' ' '-tetraacetic acid, DOTA, NSC 681107, Tetraxetan
IUPAC Name 2-[4,7,10-tris(carboxymethyl)-1,4,7,10-tetrazacyclododec-1-yl]acetic acid
SMILES OC(=O)CN1CCN(CC(=O)O)CCN(CC(=O)O)CCN(CC(=O)O)CC1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C16H28N4O8/c21-13(22)9-17-1-2-18(10-14(23)24)5-6-20(12-16(27)28)8-7-19(4-3-17)11-15(25)26/h1-12H2,(H,21,22)(H,23,24)(H,25,26)(H,27,28)
Formula Weight 404.1907
Percent Purity >95%
Chemical Name or Material DOTA
Grade HPLC