missing translation for 'onlineSavingsMsg'
Learn More

Dorzolamide hydrochloride, MedChemExpress

Product Code. 16474237
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474237 50 mg 50mg
16448348 10 mg 10mg
16418106 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474237 Supplier Medchem Express Supplier No. HYB0109A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Dorzolamide (L671152) hydrochloride is a potent carbonic anhydrase II inhibitor, with IC50 values of 0.18 nM and 600 nM for red blood cell CA-II and CA-I respectively. Dorzolamide possesses anti-tumor activity.

Dorzolamide (L671152) hydrochloride is a potent carbonic anhydrase II inhibitor, with IC50 values of 0.18 nM and 600 nM for red blood cell CA-II and CA-I respectively. Dorzolamide possesses anti-tumor activity.

Specifications

Chemical Name or Material Dorzolamide hydrochloride
CAS 130693-82-2
Color White
Purity Grade Notes Research
Molecular Formula C10H17ClN2O4S3
Quantity 50 mg
Synonym L671152 hydrochloride MK507 hydrochloride
Solubility Information DMSO : 100 mg/mL (277.09 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (34.64 mM; Need ultrasonic)
SMILES O=S(C(S1)=CC2=C1S([C@@H](C)C[C@@H]2NCC)(=O)=O)(N)=O.Cl
Molecular Weight (g/mol) 360.9
Formula Weight 360.9
Percent Purity 99.91%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.