Learn More
Dorzolamide Hydrochloride, LoGiCal
Product Code. 30459790
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459790 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459790 Supplier LoGiCal Supplier No. LGCFOR0403.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dorzolamide Hydrochloride
CAS 130693-82-2
Analyte or Component Names Dorzolamide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C10 H16 N2 O4 S3 . Cl H
Certifications/Compliance ISO 9001
Synonym Dorzolamide Hydrochloride
InChI Formula InChI=1S/C10H16N2O4S3.ClH/c1-3-12-8-4-6(2)18(13,14)10-7(8)5-9(17-10)19(11,15)16;/h5-6,8,12H,3-4H2,1-2H3,(H2,11,15,16);1H/t6-,8-;/m0./s1
Solution Type Neat
SMILES Cl.CCN[C@H]1C[C@H](C)S(=O)(=O)c2sc(cc12)S(=O)(=O)N
IUPAC Name (4S,6S)-4-(ethylamino)-6-methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 360.9
Quantity 10 mg
Formula Weight 360.0039 g/mol