Learn More
Dolutegravir sodium, MedChemExpress
Product Code. 16465578
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465578 10 mg 10mg
16448277 5 mg 5mg
16446126 100 mg 100mg
16431747 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16465578 Supplier Medchem Express Supplier No. HY13238A
 Restricted Item
Estimated Shipment: 28-09-2026
MedChemExpress Dolutegravir sodium (S/GSK1349572 sodium) is a highly potent and orally bioavailable HIV integrase strand transfer inhibitor with an IC50 of 2.7 nM for HIV-1 integrase-catalyzed strand transfer.
Dolutegravir sodium (S/GSK1349572 sodium) inhibits HIV-1 viral replication with an IC50 of 0.51 nM in peripheral blood mononuclear cells. Dolutegravir sodium (S/GSK1349572 sodium) retains a high potency against the HIV-1 Y143R, N155H, and G140S/Q148H mutants (EC50=3.6-5.8 nM).
Specifications
Chemical Name or Material Dolutegravir sodium
Physical Form Solid
CAS 1051375-19-9
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H18F2N3NaO5
For Use With (Application) COVID-19-anti-virus
Synonym S/GSK1349572 sodium
Solubility Information DMSO : ≥ 4.5 mg/mL (10.20 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CN(C2=C(O[Na])C1=O)C[C@]3([H])OCC[C@@H](C)N3C2=O)NCC4=CC=C(F)C=C4F
Molecular Weight (g/mol) 441.36
Formula Weight 441.36
Percent Purity 99.88%
Grade Research