missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl 3,4,5-Trihydroxybenzoate, Sigma-Aldrich™

Product Code. 15801061
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15801061 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15801061 Supplier Sigma Aldrich Supplier No. 8205980100
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 1166-52-5
Melting Point 94°C to 97°C
Quantity 100 g
MDL Number MFCD00002195
Synonym Dodecyl 3,4,5-trihydroxybenzoate, Dodecyl gallate, Lauryl gallate, Gallic acid dodecyl ester, Gallic acid lauryl ester, Dodecyl 3,4,5-trihydroxybenzoate
Molecular Weight (g/mol) 338.44
Chemical Name or Material Dodecyl 3,4,5-Trihydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.