Learn More
Dodecane, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 112-40-3 |
| Molecular Formula | C12H26 |
| MDL Number | MFCD00008969 |
| Quantity | 25 g |
| SMILES | CCCCCCCCCCCC |
| Molecular Weight (g/mol) | 170.33 |
| Percent Purity | 97% |
| Chemical Name or Material | Dodecane |