missing translation for 'onlineSavingsMsg'
Learn More

DOCK1 Antibody [DyLight 350], Novus Biologicals™

Product Code. 30738907 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738907 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738907 Supplier Novus Biologicals Supplier No. AF7070D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

DOCK1 Polyclonal antibody specifically detects DOCK1 in Human,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOCK1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias 180 kDa protein downstream of CRK, ced5, dedicator of cytokinesis 1, dedicator of cyto-kinesis 1, DOCK180dedicator of cytokinesis protein 1, DOwnstream of CrK
Host Species Goat
Immunogen E. coli-derived recombinant human DOCK1, Gly422-Ser666, Accession # Q14185
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 1793
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.