missing translation for 'onlineSavingsMsg'
Learn More

DNAJC6/AUXILIN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16831495 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16831495 20 μL 20µL
16821495 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16831495 Supplier Proteintech Supplier No. 219411AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNAJC6 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus, a glycine/phenylalanine (G/F)-rich region, and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain.

Specifications

Antigen DNAJC6/AUXILIN
Applications Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DNAJC6
Gene Accession No. O75061, Q80TZ3
Gene Alias AUXILIN, DJC6, DNAJC6, KIAA0473
Gene Symbols Dnajc6
Host Species Rabbit
Immunogen DNAJC6/AUXILIN Fusion Protein Ag16612
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313409, 72685, 9829
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.