missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAJB6 Monoclonal Antibody (K24018_13E3), Invitrogen™

Product Code. 30202096
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30202096 50 μL 50µg
30203637 200 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30202096 Supplier Invitrogen™ Supplier No. MA555930

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: QRHASPEDIK KAYRKLALKW HPDKNPENKE EAERKFKQVA EAYEVLSDAK KRDIYDKYGK EGLNGGGGGG SHFDSPFEFG FTFRNPDDVF REFFGGRDPF SFDFFEDPFE DFFGNRRGPR GSRSRGTGSF FSAFSGFPSF GSGFSSFDTG FTSFGSLGHG GLTSFSSTSF GGSGMGNFKS ISTSTKMVNG RKITTKRIVE NGQERVEVEE DGQLKSLTIN GVADDDALAE

This gene encodes a member of the DNAJ protein family. DNAJ family members are characterized by a highly conserved amino acid stretch called the 'J-domain' and function as one of the two major classes of molecular chaperones involved in a wide range of cellular events, such as protein folding and oligomeric protein complex assembly. This family member may also play a role in polyglutamine aggregation in specific neurons. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB6
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K24018_13E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene DNAJB6
Gene Accession No. O75190
Gene Alias DJ4; DnaJ; DnaJ (Hsp40) homolog, subfamily B, member 6; DnaJ heat shock protein family (Hsp40) member B6; dnaJ homolog subfamily B member 6; DNAJB6; dnajb6 {ECO:0000312; DnaJ-like 2 protein; EMBL:AAH78908.1, ECO:0000312; Heat shock protein J2; HHDJ1; Hsj2; HSJ-2; Hsp40 homolog; LGMD1D; LGMD1E; mammalian relative of DnaJ; mDj4; MRJ; MSJ1; MSJ-1; RGD:1308207}
Gene Symbols DNAJB6
Host Species Mouse
Immunogen Recombinant human DNAJB6 (immunogen range is 11-240).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10049
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.