missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAH17 Mouse anti-Human, Clone: 981717, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB9657-SP
This item is not returnable.
View return policy
Description
DNAH17 Monoclonal specifically detects DNAH17 in Human samples. It is validated for Immunohistochemistry.Specifications
| DNAH17 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| axonemal dynein heavy chain, axonemal dynein heavy chain-like protein 1, ciliary dynein heavy chain 17, ciliary dynein heavy chain-like protein 1, DNAHL1, DNEL2, dynein heavy chain 17, axonemal, dynein light chain 2, axonemal, dynein, axonemal, heavy chain 17, dynein, axonemal, heavy chain like 1, dynein, axonemal, heavy like 1, dynein, axonemal, heavy polypeptide 17, FLJ40457, MGC132767, MGC138489 | |
| Synthetic peptide containing human DNAH17 Accession # Q9UFH2 | |
| 25 μg | |
| 8632 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry | |
| 981717 | |
| Immunohistochemistry 0.5-25 ug/mL | |
| Q9UFH2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human DNAH17 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?