Learn More
MP Biomedicals™ Q-Bio Taq DNA polymerase
Product Code. 11477000 Shop All MP Biomedicals Products
Change view
Click to view available options
Operating Temperature: 98°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11477000 Operating Temperature: 98°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11477000 Supplier MP Biomedicals™ Supplier No. 11EPQBT025
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MP Biomedicals™ Q-Bio Taq DNA polymerase is a thermostable DNA polymerase designed for mutliplex PCR and RAPDs.
  • Q-Bio Taq DNA Polymerase is encoded by a modified form of the Thermus aquaticus DNA Polymerase gene with a N-terminal deletion.
  • No 5′-3′ exonuclease activity
  • Q-Bio Taq DNA Polymerase lacks the 5′exonuclease activity, which makes this enzyme particularly suitable for Multiplex PCR, by using several pairs of primers.
  • Highly thermostable
  • Q-Bio Taq DNA Polymerase is more thermostable than full-length Taq DNA Polymerase, as repeated exposure to 98°C does not diminish the enzyme activity.
  • Cannot degrade the 5′ end of primers
  • Q-Bio Taq DNA Polymerase has not the ability to degrade the 5′ end of primers. This is why this enzyme is particularly suitable for Multiplex PCR and RAPDs.
Specifications
Description Highly thermostable
For Use With (Application) Multiplex PCR and RAPD
Optimal Reaction Temperature 98°C