Learn More
Gibco™ DMEM/F-12, powder
Product Code. 11504466 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 L
50 L
Unit Size:
10 L
50 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11504466 50 L 50 L
11594456 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11504466 Supplier Gibco™ Supplier No. 32500043
 Restricted Item
In Stock
DMEM/F-12, powder
Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM / F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of Gibco™ DMEM / F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool. Gibco™ DMEM / F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DMEM / F-12 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DMEM / F-12 product made in our Grand Island facility (12500-096). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM / F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM / F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM / F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
TRUSTED_SUSTAINABILITY
Specifications
Form Powder
Product Type DMEM/F-12 Medium
With Additives Glutamine, Phenol Red, Sodium Pyruvate
Without Additives No HEPES, No Sodium Bicarbonate
For Use With (Application) Mammalian cell culture
Quantity 50 L
Certifications/Compliance ISO 13485 standards, FDA
pH 6.2 to 6.6
Regulatory Status CE-IVD
Content And Storage 2 to 8°C, room temperature
Description DMEM/F-12, 50L, Powder, 6.2 to 6.6 pH, 250 to 290mOsm/kg Osmolality, 36 Months Shelf Life, Ambient Shipping Condition, 2 to 8°C Storage Requirements, Animal Origin-Free Classification, For Mammalian Cell Culture