Learn More
DMAB-anabaseine Dihydrochloride, TRC
Product Code. 30010569
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010569 1 mg 1mg
30004337 10 mg 10mg
30003630 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010569 Supplier Toronto Research Chemicals Supplier No. TRCD6784081MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 154149-38-9
Molecular Formula C19 H21 N3 . 2[H Cl]
Molecular Weight (g/mol) 364.31
Synonym 4-[(5,6-Dihydro[2,3'-bipyridin]-3(4H)-ylidene)methyl]-N,N-dimethylbenzenamine Dihydrochloride;
IUPAC Name N,N-dimethyl-4-[(E)-[6-(3-pyridyl)-3,4-dihydro-2H-pyridin-5-ylidene]methyl]aniline;dihydrochloride
SMILES Cl.Cl.CN(C)c1ccc(\C=C\2/CCCN=C2c3cccnc3)cc1
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C19H21N3.2ClH/c1-22(2)18-9-7-15(8-10-18)13-16-5-4-12-21-19(16)17-6-3-11-20-14-17;;/h3,6-11,13-14H,4-5,12H2,1-2H3;2*1H/b16-13+;;
Formula Weight 363.1269
Chemical Name or Material DMAB-anabaseine Dihydrochloride